Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ndfip1
Entrez Gene ID
613097
Gene Name
Nedd4 family interacting protein 1
Gene Aliases
n4wbp5
Chromosome Location
Chr.3: 28079848 - 28113715 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032334.1 NP_001027505.1 5 - 6 564 84
XM_012959155.2 XP_012814609.1 6 - 7 786 84
XM_018092127.1 XP_017947616.1 5 - 6 883 84
GenBank mRNA BC098082.1 - 5 - 6 564 84
CU075829.1 - 5 - 6 532 84

View Full Product Details