Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
xkr5
Entrez Gene ID
619358
Gene Name
X-linked Kx blood group related 5
Gene Aliases
xrg5
Chromosome Location
Chr.Un|NW_016683369: 875590 - 888138 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033942.2 NP_001029114.1 2 - 3 321 58
XM_012964043.2 XP_012819497.1 2 - 3 356 58
GenBank mRNA AY699708.1 - 2 - 3 324 58

View Full Product Details