Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dusp1
Entrez Gene ID
448043
Gene Name
dual specificity phosphatase 1
Gene Aliases
XCL100, cl100, hvh1, mkp-1, mkp1, ptpn10
Chromosome Location
Chr.3: 32213002 - 32216490 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005450.2 NP_001005450.2 3 - 4 948 89
GenBank mRNA BC074564.1 - 3 - 4 825 89
CR760962.2 - 3 - 4 948 89

View Full Product Details