Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp1r14a
Entrez Gene ID
493331
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14A
Gene Aliases
cpi-17, ppp1inl
Chromosome Location
Chr.8: 56005494 - 56023760 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007955.2 NP_001007956.1 1 - 2 616 83
GenBank mRNA BC080468.1 - 1 - 2 464 83
CR848110.2 - 1 - 2 616 83

View Full Product Details