Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pcmtd2
Entrez Gene ID
496500
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.10: 575445 - 582838 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011087.2 NP_001011087.2 3 - 4 490 87
GenBank mRNA CR761105.1 - 3 - 4 490 87

View Full Product Details