Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
slc27a6
Entrez Gene ID
496813
Gene Name
solute carrier family 27 member 6
Gene Aliases
acsvl2, facvl2, fatp6, slc27a2, vlcs-h1
Chromosome Location
Chr.1: 158907413 - 158922075 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011348.2 NP_001011348.2 1 - 2 835 88
GenBank mRNA CR848474.2 - 1 - 2 835 88

View Full Product Details