Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arl10b
Entrez Gene ID
548865
Gene Name
ADP-ribosylation factor-like 10B
Gene Aliases
-
Chromosome Location
Chr.3: 60848266 - 60870465 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016111.2 NP_001016111.1 6 - 7 638 104
GenBank mRNA BC155533.1 - 6 - 7 673 104
CR762112.2 - 6 - 7 638 104

View Full Product Details