Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kif11
Entrez Gene ID
548870
Gene Name
kinesin family member 11
Gene Aliases
Cos2, Costal2, eg5, hksp, kif11-a, kif11-b, knsl1, trip5
Chromosome Location
Chr.7: 17322128 - 17341973 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016116.3 NP_001016116.2 10 - 11 1309 90
XM_012966561.1 XP_012822015.1 11 - 12 1469 90
GenBank mRNA BC166147.1 - 10 - 11 1240 90
CR762094.2 - 10 - 11 1309 90
EF524557.1 - 9 - 10 1128 90

View Full Product Details