Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
alg12
Entrez Gene ID
548874
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
ecm39, halg12, pp14673
Chromosome Location
Chr.3: 62982647 - 62995575 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016120.2 NP_001016120.1 3 - 4 678 99
GenBank mRNA BC121879.1 - 3 - 4 959 99
BC155512.1 - 3 - 4 920 99
CR762085.2 - 3 - 4 678 99

View Full Product Details