Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gins3
Entrez Gene ID
548877
Gene Name
GINS complex subunit 3 (Psf3 homolog)
Gene Aliases
psf3
Chromosome Location
Chr.4: 27782952 - 27787756 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016123.2 NP_001016123.1 1 - 2 176 58
GenBank mRNA BC159002.1 - 1 - 2 228 58
CR762072.2 - 1 - 2 176 58

View Full Product Details