Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
paqr3
Entrez Gene ID
548889
Gene Name
progestin and adipoQ receptor family member 3
Gene Aliases
-
Chromosome Location
Chr.1: 75963458 - 75981263 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012962038.2 XP_012817492.1 7 - 8 1117 81
XM_012962044.2 XP_012817498.1 7 - 8 991 81
XM_012962048.2 XP_012817502.1 6 - 7 1074 81
GenBank mRNA BC171300.1 - 6 - 7 878 81
CR762035.2 - 6 - 7 879 81

View Full Product Details