Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gins1
Entrez Gene ID
548918
Gene Name
GINS complex subunit 1 (Psf1 homolog)
Gene Aliases
psf1
Chromosome Location
Chr.5: 49107251 - 49112420 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016164.2 NP_001016164.1 2 - 3 160 84
GenBank mRNA BC168788.1 - 2 - 3 382 84
BC170763.1 - 2 - 3 160 84
BC170767.1 - 2 - 3 160 84
CR761910.2 - 2 - 3 160 84

View Full Product Details