Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cks2
Entrez Gene ID
548924
Gene Name
CDC28 protein kinase regulatory subunit 2
Gene Aliases
Xe-p9, ckshs2
Chromosome Location
Chr.1: 125658810 - 125662049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016170.2 NP_001016170.1 2 - 3 217 88
GenBank mRNA BC170572.1 - 2 - 3 216 88
BC170574.1 - 2 - 3 216 88
CR761887.2 - 2 - 3 217 88

View Full Product Details