Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
c7orf26
Entrez Gene ID
548985
Gene Name
chromosome 7 open reading frame 26
Gene Aliases
-
Chromosome Location
Chr.9: 14385044 - 14391098 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016231.2 NP_001016231.1 4 - 5 619 98
GenBank mRNA BC123054.1 - 2 - 3 259 98
BC171225.1 - 4 - 5 619 98
BC171227.1 - 4 - 5 619 98
CR926283.2 - 4 - 5 619 98

View Full Product Details