Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
angel1
Entrez Gene ID
548990
Gene Name
angel homolog 1
Gene Aliases
-
Chromosome Location
Chr.8: 61379884 - 61395244 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016236.2 NP_001016236.1 - 1328 94
GenBank mRNA BC122891.1 - - 1319 94
BC166333.1 - - 1248 94
BC171302.1 - - 1301 94
CR926268.2 - - 1328 94

View Full Product Details