Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnpy2
Entrez Gene ID
549005
Gene Name
canopy FGF signaling regulator 2
Gene Aliases
tmem4
Chromosome Location
Chr.2: 134004635 - 134019284 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016251.2 NP_001016251.1 2 - 3 164 76
GenBank mRNA BC166940.1 - 2 - 3 389 76
BC170706.1 - 1 - 2 129 76
BC170708.1 - 1 - 2 129 76
CR761752.2 - 2 - 3 164 76

View Full Product Details