Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kti12
Entrez Gene ID
549045
Gene Name
KTI12 chromatin associated homolog
Gene Aliases
sbbi81, tot4
Chromosome Location
Chr.3: 81044638 - 81072227 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016291.2 NP_001016291.1 7 - 8 587 65
XM_012966570.1 XP_012822024.1 7 - 8 781 65
XM_018092083.1 XP_017947572.1 7 - 8 620 65
GenBank mRNA BC170877.1 - 6 - 7 567 65
BC170879.1 - 6 - 7 567 65
CR761570.2 - 7 - 8 587 65

View Full Product Details