Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cox18
Entrez Gene ID
549087
Gene Name
cytochrome c oxidase assembly factor COX18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683393: 578738 - 587720 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016333.2 NP_001016333.1 1 - 2 754 89
XM_012962186.2 XP_012817640.1 2 - 3 596 89
XM_012962189.2 XP_012817643.1 2 - 3 595 89
XM_012962191.1 XP_012817645.1 2 - 3 512 89
GenBank mRNA BC171112.1 - 1 - 2 666 89
CR761413.2 - 1 - 2 754 89

View Full Product Details