Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cox18
Entrez Gene ID
549087
Gene Name
cytochrome c oxidase assembly factor COX18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683393: 578738 - 587720 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016333.2 NP_001016333.1 5 - 6 1240 79
XM_012962186.2 XP_012817640.1 6 - 7 1082 79
XM_012962189.2 XP_012817643.1 6 - 7 1081 79
XM_012962191.1 XP_012817645.1 6 - 7 998 79
XM_018089476.1 XP_017944965.1 5 - 6 748 79
GenBank mRNA BC171112.1 - 5 - 6 1152 79
CR761413.2 - 5 - 6 1240 79

View Full Product Details