Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhbdd1
Entrez Gene ID
549203
Gene Name
rhomboid domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 115215706 - 115247077 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016449.2 NP_001016449.1 4 - 5 741 103
XM_012963006.2 XP_012818460.1 4 - 5 809 103
GenBank mRNA CR760998.2 - 4 - 5 741 103

View Full Product Details