Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
npm3
Entrez Gene ID
549210
Gene Name
nucleophosmin/nucleoplasmin 3
Gene Aliases
no29, pormim, tmem123
Chromosome Location
Chr.7: 22888023 - 22906029 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016456.2 NP_001016456.1 4 - 5 488 85
GenBank mRNA BC167920.1 - 4 - 5 473 85
CR760966.2 - 4 - 5 488 85

View Full Product Details