Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
polr2e
Entrez Gene ID
549227
Gene Name
polymerase (RNA) II (DNA directed) polypeptide E, 25kDa
Gene Aliases
-
Chromosome Location
Chr.1: 81247073 - 81255347 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016473.2 NP_001016473.1 2 - 3 243 104
GenBank mRNA BC157231.1 - 3 - 4 445 104
CR760906.2 - 2 - 3 243 104

View Full Product Details