Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
polr2e
Entrez Gene ID
549227
Gene Name
polymerase (RNA) II (DNA directed) polypeptide E, 25kDa
Gene Aliases
-
Chromosome Location
Chr.1: 81247073 - 81255347 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016473.2 NP_001016473.1 4 - 5 441 83
GenBank mRNA BC157231.1 - 5 - 6 643 83
CR760906.2 - 4 - 5 441 83

View Full Product Details