Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tex30
Entrez Gene ID
549282
Gene Name
testis expressed 30
Gene Aliases
c13orf27
Chromosome Location
Chr.2: 116023290 - 116030098 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016528.2 NP_001016528.1 2 - 3 270 101
XM_012956974.2 XP_012812428.1 2 - 3 334 101
GenBank mRNA BC170894.1 - 2 - 3 259 101
BC170896.1 - 2 - 3 259 101
CR926187.2 - 2 - 3 270 101

View Full Product Details