Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pole4
Entrez Gene ID
549359
Gene Name
polymerase (DNA-directed), epsilon 4, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.3: 30696244 - 30701213 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016605.2 NP_001016605.1 1 - 2 229 69
GenBank mRNA BC170722.1 - 1 - 2 228 69
CR760539.2 - 1 - 2 229 69

View Full Product Details