Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dtwd2
Entrez Gene ID
549361
Gene Name
DTW domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 162152479 - 162207806 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016607.2 NP_001016607.1 5 - 6 606 87
GenBank mRNA BC158499.1 - 5 - 6 606 87
CR760536.2 - 5 - 6 606 87

View Full Product Details