Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkbp3
Entrez Gene ID
549377
Gene Name
FK506 binding protein 3
Gene Aliases
-
Chromosome Location
Chr.8: 71674513 - 71683388 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016623.2 NP_001016623.1 5 - 6 512 103
XM_012968431.2 XP_012823885.1 5 - 6 560 103
GenBank mRNA BC167937.1 - 5 - 6 507 103
BC170855.1 - 5 - 6 503 103
BC170857.1 - 5 - 6 505 103
CR760490.2 - 5 - 6 512 103

View Full Product Details