Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rpa3
Entrez Gene ID
549380
Gene Name
replication protein A3
Gene Aliases
repa3
Chromosome Location
Chr.Un|NW_016683410: 1466811 - 1470998 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016626.2 NP_001016626.1 3 - 4 294 95
GenBank mRNA BC158993.1 - 3 - 4 370 95
BC166941.1 - 3 - 4 310 95
CR760473.2 - 3 - 4 294 95

View Full Product Details