Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk2ap1
Entrez Gene ID
549484
Gene Name
cyclin-dependent kinase 2 associated protein 1
Gene Aliases
doc-1, doc1, dorc1, p12doc-1, st19
Chromosome Location
Chr.1: 152517891 - 152524389 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016730.2 NP_001016730.1 2 - 3 168 85
XM_018093477.1 XP_017948966.1 2 - 3 351 85
GenBank mRNA BC084911.1 - 2 - 3 271 85
BC121523.1 - 2 - 3 187 85
BC167332.1 - 2 - 3 379 85
CR760030.2 - 2 - 3 168 85

View Full Product Details