Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zbtb8os
Entrez Gene ID
549545
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
arch, arch2
Chromosome Location
Chr.2: 86190955 - 86199276 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016791.2 NP_001016791.1 1 - 2 156 67
GenBank mRNA CR855646.2 - 1 - 2 156 67

View Full Product Details