Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mri1
Entrez Gene ID
549631
Gene Name
methylthioribose-1-phosphate isomerase 1
Gene Aliases
mtna
Chromosome Location
Chr.3: 130028476 - 130036066 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016877.2 NP_001016877.1 4 - 5 777 93
GenBank mRNA BC118767.1 - 4 - 5 735 93
CR848601.2 - 4 - 5 777 93

View Full Product Details