Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsmce3
Entrez Gene ID
549684
Gene Name
NSE3 homolog, SMC5-SMC6 complex component
Gene Aliases
hca4, magee1, mageg1, magel3, ndnl2, nse3
Chromosome Location
Chr.8: 8604663 - 8611843 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016930.2 NP_001016930.1 10 - 11 790 98
XM_012954225.2 XP_012809679.1 10 - 11 896 98
XM_012954226.2 XP_012809680.1 10 - 11 823 98
GenBank mRNA BC155975.1 - 10 - 11 795 98
CR762194.2 - 10 - 11 790 98

View Full Product Details