Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rnf5
Entrez Gene ID
549825
Gene Name
ring finger protein 5
Gene Aliases
ring5, rma1, rnf185
Chromosome Location
Chr.8: 54329606 - 54340733 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017071.2 NP_001017071.1 2 - 3 132 106
GenBank mRNA CR761547.2 - 2 - 3 132 106

View Full Product Details