Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taldo1
Entrez Gene ID
549885
Gene Name
transaldolase 1
Gene Aliases
-
Chromosome Location
Chr.4: 36207979 - 36223827 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017131.2 NP_001017131.1 4 - 5 637 73
GenBank mRNA CR761089.2 - 4 - 5 637 73

Gene Symbol
LOC105947067
Entrez Gene ID
105947067
Gene Name
transaldolase-like
Gene Aliases
-
Chromosome Location
Chr.4: 38091513 - 38098994 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018093172.1 XP_017948661.1 3 - 4 373 73

Gene Symbol
LOC108644509
Entrez Gene ID
108644509
Gene Name
transaldolase-like
Gene Aliases
-
Chromosome Location
Chr.4: 18227649 - 18228476 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018093353.1 XP_017948842.1 1 - 2 206 73

Gene Symbol
LOC108644521
Entrez Gene ID
108644521
Gene Name
transaldolase-like
Gene Aliases
-
Chromosome Location
Chr.4: 18248194 - 18253128 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018093355.1 XP_017948844.1 2 - 3 261 73

Gene Symbol
LOC108647424
Entrez Gene ID
108647424
Gene Name
transaldolase-like
Gene Aliases
-
Chromosome Location
Chr.4: 38055651 - 38056483 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018093403.1 XP_017948892.1 1 - 2 283 73

View Full Product Details