Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sap30l
Entrez Gene ID
549895
Gene Name
SAP30-like
Gene Aliases
ns4atp2, sap30l-a, sap30l-b
Chromosome Location
Chr.3: 9199327 - 9209454 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017141.2 NP_001017141.1 3 - 4 432 66
XM_012959085.2 XP_012814539.1 4 - 5 1525 66
GenBank mRNA CR760983.2 - 3 - 4 432 66

View Full Product Details