Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfand3
Entrez Gene ID
549901
Gene Name
zinc finger AN1-type containing 3
Gene Aliases
tex27
Chromosome Location
Chr.Un|NW_016683574: 83538 - 122754 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017147.2 NP_001017147.1 - 197 91
GenBank mRNA BC157766.1 - - 241 91
CR760946.2 - - 197 91

View Full Product Details