Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cirbp
Entrez Gene ID
549982
Gene Name
cold inducible RNA binding protein
Gene Aliases
CIRP2, Xcirp, cirbp-a, cirbp-b, cirbp2, cirp, cirp1, xCIRP2
Chromosome Location
Chr.1: 80999043 - 81005134 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017228.3 NP_001017228.1 2 - 3 171 99
GenBank mRNA CF782242.1 - 2 - 3 193 99
CR760272.2 - 2 - 3 156 99

View Full Product Details