Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taf10
Entrez Gene ID
550033
Gene Name
TATA-box binding protein associated factor 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683661: 50 - 995 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017279.2 NP_001017279.1 - 334 64
GenBank mRNA CR855741.2 - - 334 64

View Full Product Details