Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
clns1a
Entrez Gene ID
496644
Gene Name
chloride channel, nucleotide-sensitive, 1A
Gene Aliases
clci, clns1b, icln
Chromosome Location
Chr.2: 165533820 - 165545771 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037262.1 NP_001032339.1 4 - 5 772 89
GenBank mRNA BC087765.1 - 4 - 5 742 89
BC157253.1 - 4 - 5 766 89
CR761131.2 - 4 - 5 772 89

View Full Product Details