Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
c12orf10
Entrez Gene ID
594885
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
-
Chromosome Location
Chr.2: 144274621 - 144282127 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039742.1 NP_001034831.1 6 - 7 972 59
XM_012957066.2 XP_012812520.1 6 - 7 967 59
GenBank mRNA BC091097.1 - 6 - 7 968 59
CR761706.2 - 6 - 7 972 59

View Full Product Details