Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhebl1
Entrez Gene ID
394534
Gene Name
RHEB like 1
Gene Aliases
rhebl1c
Chromosome Location
Chr.2: 133166719 - 133172807 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203606.2 NP_988937.1 7 - 8 537 87
GenBank mRNA BC061397.1 - 7 - 8 560 87
CR761520.2 - 7 - 8 537 87

View Full Product Details