Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grwd1
Entrez Gene ID
394881
Gene Name
glutamate-rich WD repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 106896760 - 106903743 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203937.3 NP_989268.3 4 - 5 762 92
GenBank mRNA BC064215.1 - 4 - 5 754 92
CR760996.2 - 4 - 5 92

View Full Product Details