Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nfe2l2
Entrez Gene ID
493217
Gene Name
nuclear factor, erythroid 2 like 2
Gene Aliases
NF-E2, NF-E2R2, nrf2
Chromosome Location
Chr.9: 60561849 - 60575420 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007489.2 NP_001007490.1 4 - 5 669 77
GenBank mRNA BC079927.1 - 4 - 5 694 77
CR942453.2 - 4 - 5 669 77

View Full Product Details