Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gadd45a
Entrez Gene ID
548905
Gene Name
growth arrest and DNA damage inducible alpha
Gene Aliases
ddit1, gadd, gadd45
Chromosome Location
Chr.4: 80059638 - 80064270 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016151.3 NP_001016151.1 1 - 2 156 72
GenBank mRNA BC121532.1 - 1 - 2 156 72
CR761969.2 - 1 - 2 182 72

View Full Product Details