Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
b4galt5
Entrez Gene ID
548965
Gene Name
UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 5
Gene Aliases
-
Chromosome Location
Chr.6: 96791543 - 96816841 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016211.3 NP_001016211.1 7 - 8 1087 80
XM_012964682.1 XP_012820136.1 8 - 9 1054 80
XM_018094645.1 XP_017950134.1 7 - 8 866 80
XM_018094646.1 XP_017950135.1 7 - 8 854 80
GenBank mRNA BC122889.1 - 7 - 8 1087 80
CR926330.2 - 8 - 9 1027 80

View Full Product Details