Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
naxd
Entrez Gene ID
549064
Gene Name
NAD(P)HX dehydratase
Gene Aliases
carkd
Chromosome Location
Chr.2: 113588482 - 113605453 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016310.4 NP_001016310.2 6 - 7 1476 70
NM_001247985.1 NP_001234914.1 5 - 6 488 70
XM_012956947.2 XP_012812401.1 5 - 6 589 70
XM_012956948.2 XP_012812402.1 5 - 6 679 70
XM_018091246.1 XP_017946735.1 6 - 7 3426 70
XM_018091247.1 XP_017946736.1 5 - 6 3059 70
GenBank mRNA BC121541.1 - 6 - 7 1436 70
CR761491.2 - 6 - 7 904 70
CX372587.2 - 5 - 6 413 70

View Full Product Details