Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc1d7
Entrez Gene ID
549086
Gene Name
TBC1 domain family member 7
Gene Aliases
pig51
Chromosome Location
Chr.6: 84569069 - 84575250 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016332.3 NP_001016332.2 4 - 5 676 103
XM_012964694.2 XP_012820148.1 3 - 4 474 103
GenBank mRNA BC121213.1 - 4 - 5 676 103
CR761423.2 - 3 - 4 474 103

View Full Product Details