Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exd2
Entrez Gene ID
549208
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
exdl2
Chromosome Location
Chr.8: 89460401 - 89477717 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016454.3 NP_001016454.2 5 - 6 1198 91
XM_012968412.2 XP_012823866.1 6 - 7 1302 91
XM_012968413.2 XP_012823867.1 6 - 7 1150 91
XM_018096255.1 XP_017951744.1 6 - 7 1177 91
GenBank mRNA BC124061.1 - 5 - 6 1198 91

View Full Product Details