Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
znhit1
Entrez Gene ID
549810
Gene Name
zinc finger, HIT-type containing 1
Gene Aliases
cg1i, znfn4a1
Chromosome Location
Chr.3: 136818260 - 136820515 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017056.3 NP_001017056.2 2 - 3 502 59
GenBank mRNA BC121538.1 - 2 - 3 502 59

View Full Product Details