Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ggct
Entrez Gene ID
613056
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
-
Chromosome Location
Chr.6: 72882557 - 72902869 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044400.1 NP_001037865.1 3 - 4 450 91
XM_012964807.2 XP_012820261.1 4 - 5 631 91
GenBank mRNA BC096512.1 - 3 - 4 411 91
BC121204.1 - 3 - 4 421 91
BC161542.1 - 3 - 4 430 91
CR942442.2 - 3 - 4 450 91

View Full Product Details